Learn More
STK39 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STK39.
Brand: Abnova PAB3097.400uL
Description
This gene encodes a serine/threonine kinase that is thought to function in the cellular stress response pathway. The kinase is activated in response to hypotonic stress, leading to phosphorylation of several cation-chloride-coupled cotransporters. The catalytically active kinase specifically activates the p38 MAP kinase pathway, and its interaction with p38 decreases upon cellular stress, suggesting that this kinase may serve as an intermediate in the response to cellular stress. [provided by RefSeq
Specifications
| STK39 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of STK39. | |
| In PBS (0.09% sodium azide) | |
| DCHT/DKFZp686K05124/PASK/SPAK | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 27347 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| STK39 | |
| STK39 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human STK39. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only