Learn More
STK19, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STK19.
Brand: Abnova PAB2043.400uL
Description
This gene encodes a serine/threonine kinase which localizes predominantly to the nucleus. Its specific function is unknown; it is possible that phosphorylation of this protein is involved in transcriptional regulation. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6 and expresses two transcript variants. [provided by RefSeq
Specifications
| STK19 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of STK19. | |
| In PBS (0.09% sodium azide) | |
| D6S60/D6S60E/G11/HLA-RP1/MGC117388/RP1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 8859 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| STK19 | |
| STK19 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human STK19. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only