Learn More
STIM2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STIM2.
Brand: Abnova PAB13266.100ug
Description
This gene is a member of the stromal interaction molecule (STIM) family and likely arose, with related family member STIM1, from a common ancestral gene. It encodes a type 1 transmembrane protein whose function is currently unknown, but is a likely adhesion molecule with a role in early hematopoiesis. Alternative translation initiation from an AUG and a non-AUG (UUG) start site results in the production of two different isoforms. [provided by RefSeq
Specifications
| STIM2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of STIM2. | |
| In PBS (0.02% sodium azide) | |
| FLJ39527/KIAA1482 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| STIM2 | |
| STIM2 | |
| A synthetic peptide corresponding to internal region 17 amino acids of human STIM2. | |
| RUO | |
| 57620 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only