missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5B Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a synthetic peptide of Stat5b.
Brand: Abnova PAB0835.200ug
This item is not returnable.
View return policy
Specifications
| STAT5B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a synthetic peptide of Stat5b. | |
| In PBS (0.2% gelatin, 0.09% sodium azide) | |
| Stat5b | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of mouse Stat5b. | |
| RUO | |
| 20851 | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:200) The optimal working dilution should be determined by the end user. | |
| Stat5b | |
| Rabbit | |
| 200 μg | |
| Primary | |
| This antibody reacts with STAT5a and STAT5a/b. | |
| Store at 4°C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction