Learn More
STAT5A (phospho S726), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of STAT5A.
Brand: Abnova PAB0620.400uL
Description
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is activated by, and mediates the responses of many cell ligands, such as IL2, IL3, IL7 GM-CSF, erythropoietin, thrombopoietin, and different growth hormones. Activation of this protein in myeloma and lymphoma associated with a TEL/JAK2 gene fusion is independent of cell stimulus and has been shown to be essential for the tumorigenesis. The mouse counterpart of this gene is found to induce the expression of BCL2L1/BCL-X(L), which suggests the antiapoptotic function of this gene in cells. [provided by RefSeq
Specifications
| STAT5A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of STAT5A. | |
| In PBS (0.09% sodium azide) | |
| MGF/STAT5 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 6776 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
| STAT5A | |
| STAT5A | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S726 of human STAT5A. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only