Learn More
STAP2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant STAP2.
Brand: Abnova PAB21900.100uL
Description
This gene encodes the substrate of breast tumor kinase, an Src-type nonreceptor tyrosine kinase. The encoded protein possesses domains and several tyrosine phosphorylation sites characteristic of adaptor proteins that mediate the interactions linking proteins involved in signal transduction pathways. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: QGLTIYFYNSNRDFQHVEKLNLGAFEKLTDEIPWGSSRDPGTHFSLILRNQEIKFKVETLECREMWKGFILTVVELRVPTDLTLLPGHLYMMSEVLAKEEARRALETPSCFLKVSRLEAQLLLESpecifications
| STAP2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant STAP2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| BKS/FLJ20234 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55620 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| STAP2 | |
| STAP2 | |
| Recombinant protein corresponding to amino acids of human STAP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only