Learn More
ST8SIA6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ST8SIA6.
Brand: Abnova PAB20556.100uL
Description
Sialic acid is a key determinate of oligosaccharide structures involved in cell-cell communication, cell-substrate interaction, adhesion, and protein targeting. ST8SIA6 belongs to a family of sialyltransferases (EC 2.4.99.8) that synthesize sialylglycoconjugates (Takashima et al., 2002 [PubMed 11980897]).[supplied by OMIM
Sequence: RILVEESREATHGTPAALRTLRSPATAVPRATNSTYLNEKSLQLTEKCKNLQYGIESFSNKTKGYSENDYLQIITDIQSCPWKRQAEEYANFRAKLASCCDAVQNFVVSQNNTPVGTNMSYEVESKKEIPIKKNIFHMFPVSQPSpecifications
| ST8SIA6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ST8SIA6. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| SIA8F/SIAT8F/ST8SIA-VI | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 338596 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| ST8SIA6 | |
| ST8SIA6 | |
| Recombinant protein corresponding to amino acids of human ST8SIA6. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only