Learn More
ST3GAL6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ST3GAL6
Brand: Abnova PAB28018.100uL
Description
Sialyltransferases, such as ST3GAL6, catalyze the transfer of sialic acid from cytidine 5-prime monophospho-N-acetylneuraminic acid (CMP-NeuAc) to terminal positions of glycoprotein and glycolipid carbohydrate groups. Terminal NeuAc residues are key determinants of carbohydrate structures, such as the sialyl-Lewis X determinants, and are widely distributed in many cell types.[supplied by OMIM
Sequence: IQPCLSKPAFASLLRFHQFHPFLCAADFRKIASLYGSDKFDLPYGMRTSAEYFRLALSKLQSCDLFDEFDNIPCKKCVVVGNGGVLKNKTLGEKIDSYDVIIRMNNGSpecifications
| ST3GAL6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ST3GAL6 | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| SIAT10/ST3GALVI | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10402 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50 - 1:200) Western Blot (1:100 - 1:250) The optimal working dilution should be determined by the end user. | |
| ST3GAL6 | |
| ST3GAL6 | |
| Recombinant protein corresponding to amino acids of human ST3GAL6 | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only