Learn More
SSTR5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SSTR5.
Brand: Abnova PAB26474.50ug
Description
Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The biological effects of somatostatin are probably mediated by a family of G protein-coupled receptors that are expressed in a tissue-specific manner. SSTR5 is a member of the superfamily of receptors having seven transmembrane segments. [provided by RefSeq
Specifications
| SSTR5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SSTR5. | |
| In PBS (0.09% sodium azide) | |
| SSTR5 | |
| A synthetic peptide corresponding to 17 amino acids at C-terminus of human SSTR5. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (23 ug/mL) The optimal working dilution should be determined by the end user. | |
| SSTR5 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 6755 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only