Learn More
SSR3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SSR3.
Brand: Abnova PAB20781.100uL
Description
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR is comprised of four membrane proteins/subunits: alpha, beta, gamma, and delta. The first two are glycosylated subunits and the latter two are nonglycosylated subunits. The protein encoded by this gene is the gamma subunit and is predicted to span the membrane four times. [provided by RefSeq
Sequence: VLKHKVAQKREDAVSKEVTRKLSEADNRKMSRKEKDERILWKKNEVADYEASpecifications
| SSR3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SSR3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| TRAPG | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6747 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| SSR3 | |
| SSR3 | |
| Recombinant protein corresponding to amino acids of human SSR3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only