Learn More
SPEN, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SPEN.
Brand: Abnova PAB20849.100uL
Description
This gene encodes a hormone inducible transcriptional repressor. Repression of transcription by this gene product can occur through interactions with other repressors, by the recruitment of proteins involved in histone deacetylation, or through sequestration of transcriptional activators. The product of this gene contains a carboxy-terminal domain that permits binding to other corepressor proteins. This domain also permits interaction with members of the NuRD complex, a nucleosome remodeling protein complex that contains deacetylase activity. In addition, this repressor contains several RNA recognition motifs that confer binding to a steroid receptor RNA coactivator; this binding can modulate the activity of both liganded and nonliganded steroid receptors. [provided by RefSeq
Sequence: NRERAYEHSAYGHHERGTGGFDRTRHYDQDYYRDPRERTLQHGLYYASRSRSPNRFDAHDPRYEPRAREQFTLPSVVHRDIYRDDITREVRGRRPERNYQHSRSRSPHSSQSRNQSPQRLASQASRPTRSPSGSGSRSRSSSSDSISpecifications
| SPEN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SPEN. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| KIAA0929/MINT/RP1-134O19.1/SHARP | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23013 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| SPEN | |
| SPEN | |
| Recombinant protein corresponding to amino acids of human SPEN. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only