Learn More
SPARC Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SPARC.
Brand: Abnova PAB28632.100uL
Description
Secreted protein acidic and rich in cysteine/osteonectin/BM40, or SPARC, is a matrix-associated protein that elicits changes in cell shape, inhibits cell-cycle progression, and influences the synthesis of extracellular matrix (ECM) (Bradshaw et al., 2003 [PubMed 12721366]).[supplied by OMIM
Sequence: QEALPDETEVVEETVAEVTEVSVGANPVQVEVGEFDDGAEETEEEVVAENPCQNHHCKHGKVCELDENNTPMCVCQDPTSCPAPIGEFEKVCSNDNKTFDSSCHFFATKCTLEGTKKGHKLHLDYSpecifications
| SPARC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SPARC. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| ON | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6678 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| SPARC | |
| SPARC | |
| Recombinant protein corresponding to amino acids of human SPARC. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only