Learn More
SOX3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SOX3.
Brand: Abnova PAB14857.100ug
Description
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene have been associated with X-linked mental retardation with growth hormone deficiency. [provided by RefSeq
Specifications
| SOX3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SOX3. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| GHDX/MRGH/PHP/PHPX/SOXB | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 6658 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| SOX3 | |
| SOX3 | |
| A synthetic peptide corresponding to N-terminus of human SOX3. | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼46 KDa of human SOX3. | |
| Store at 4°C. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only