missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sod2, Rabbit, R-PE, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Recombinant of Sod2.
Brand: Abnova PAB28998.100ug
This item is not returnable.
View return policy
Specifications
| Sod2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Recombinant of Sod2. | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Sod2 | |
| Recombinant protien corresponding rat Sod2. | |
| 100 μg | |
| Primary | |
| Detects a ∼25 kDa protein corresponding to the molecular mass of Mn superoxide dismutase (SOD) on SDS PAGE immunoblots. | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| R-PE | |
| ELISA Immunofluorescence Immunohistochemistry Immunoprecipitation Western blot (0.5 ug/ml) The optimal working dilution should be determined by the end user. | |
| Sod2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 24787 | |
| Bovine, Canine, Chicken, Drosophila, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus species | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction