Learn More
SOD1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SOD1.
Brand: Abnova PAB18299.100ug
Description
The protein encoded by this gene binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. The encoded isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene. [provided by RefSeq
Specifications
| SOD1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(1 ug/mL) Immunohistochemistry (Frozen sections)(1 ug/mL) Immunocytochemistry (1 ug/mL) The optimal working dilution should be determined by the end use | |
| SOD1 | |
| SOD1 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human SOD1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
| Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of SOD1. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| ALS/ALS1/IPOA/SOD/homodimer | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 6647 | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only