missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SOCS1.
Brand: Abnova PAB12778.100ug
This item is not returnable.
View return policy
Specifications
| SOCS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SOCS1. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| SOCS1 | |
| A synthetic peptide corresponding to C-terminus of human and mouse SOCS1. | |
| RUO | |
| 8651/12703 | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| CIS1/CISH1/JAB/SOCS-1/SSI-1/SSI1/TIP3 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼24 KDa of SOCS1. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction