Learn More
SNX18, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SNX18.
Brand: Abnova PAB23168.100uL
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a SH3 domain. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: DDEWDDSSTVADEPGALGSGAYPDLDGSSSAGVGAAGRYRLSTRSDLSLGSRGGSVPPQHHPSGPKSSATVSRNLNSpecifications
| SNX18 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SNX18. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ11997/FLJ32560/MGC150827/MGC150829/SH3PX2/SH3PXD3B/SNAG1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 112574 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| SNX18 | |
| SNX18 | |
| Recombinant protein corresponding to amino acids of human SNX18. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only