Learn More
SNW1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SNW1.
Marca: Abnova PAB28666.100uL
Descrizione
This gene, a member of the SNW gene family, encodes a coactivator that enhances transcription from some Pol II promoters. This coactivator can bind to the ligand-binding domain of the vitamin D receptor and to retinoid receptors to enhance vitamin D-, retinoic acid-, estrogen-, and glucocorticoid-mediated gene expression. It can also function as a splicing factor by interacting with poly(A)-binding protein 2 to directly control the expression of muscle-specific genes at the transcriptional level. Finally, the protein may be involved in oncogenesis since it interacts with a region of SKI oncoproteins that is required for transforming activity. [provided by RefSeq
Sequence: RNLSRAAPDKRSKLQRNENRDISEVIALGVPNPRTSNEVQYDQRLFNQSKGMDSGFAGGEDEIYNVYDQAWRGGKDMAQSIYRPSKNLDKDMYGDDLEARIKTNRFVPDKEFSGSDRRQRGREGPVQFEEDPFGLDKFSpecifica
| SNW1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SNW1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| Bx42/MGC119379/NCOA-62/PRPF45/Prp45/SKIIP/SKIP | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 22938 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(1:50-1:200) Immunofluorescence (1-4 ug/ml) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| SNW1 | |
| SNW1 | |
| Recombinant protein corresponding to amino acids of human SNW1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only