Learn More
SNRNP40 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SNRNP40.
Brand: Abnova PAB21803.100uL
Description
This gene encodes a component of the U5 small nuclear ribonucleoprotein (snRNP) particle. The U5 snRNP is part of the spliceosome, a multiprotein complex that catalyzes the removal of introns from pre-messenger RNAs. [provided by RefSeq
Sequence: QGNVHNFEKNLLRCSWSPDGSKIAAGSADRFVYVWDTTSRRILYKLPGHAGSINEVAFHPDEPIIISASSDKRLYMGEIQSpecifications
| SNRNP40 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SNRNP40. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| 40K/FLJ41108/HPRP8BP/MGC1910/PRP8BP/PRPF8BP/RP11-490K7.3/SPF38/WDR57 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9410 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| SNRNP40 | |
| SNRNP40 | |
| Recombinant protein corresponding to amino acids of human SNRNP40. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only