missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC1B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SMC1B.
Brand: Abnova PAB28446.100uL
This item is not returnable.
View return policy
Description
SMC1L2 belongs to a family of proteins required for chromatid cohesion and DNA recombination during meiosis and mitosis (3:Revenkova et al., 2001 [PubMed 11564881]).[supplied by OMIM
Sequence: KSTMQILSVQAEENCLQTVNELMAKQQQLKDIRVTQNSSAEKVQTQIEEERKKFLAVDREVGKLQKEVVSIQTSLEQKRLEKHNLLLDCKVQDIEIILLSGSLDDIIEVEMGTEAESTQATIDIYEKEEAFEIDYSSLSpecifications
| SMC1B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SMC1B. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| FLJ43748/SMC1BETA/SMC1L2/bK268H5/bK268H5.5 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 27127 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry(1:200-1:500) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| SMC1B | |
| SMC1B | |
| Recombinant protein corresponding to amino acids of recombinant SMC1B. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction