Learn More
SMARCD1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SMARCD1.
Brand: Abnova PAB20261.100uL
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MGPAPGQGLYRSPMPGAAYPRPGMLPGSRMTPQGPSMGPPGYGGNPSVRPGLAQSGMDQSRKRPAPQQIQQVQQQAVQNRNHNAKKSpecifications
| SMARCD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SMARCD1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| BAF60A/CRACD1/Rsc6p | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6602 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| SMARCD1 | |
| SMARCD1 | |
| Recombinant protein corresponding to amino acids of human SMARCD1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only