Learn More
SLITRK1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLITRK1.
Brand: Abnova PAB13327.100ug
Beschreibung
Members of the SLITRK family, such as SLITRK1, are integral membrane proteins with 2 N-terminal leucine-rich repeat (LRR) domains similar to those of SLIT proteins (see SLIT1; MIM 603742). Most SLITRKs, but not SLITRK1, also have C-terminal regions that share homology with neurotrophin receptors (see NTRK1; MIM 191315). SLITRKs are expressed predominantly in neural tissues and have neurite-modulating activity (Aruga et al., 2003 [PubMed 14557068]).[supplied by OMIM
Spezifikation
| SLITRK1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SLITRK1. | |
| In PBS (0.02% sodium azide) | |
| FLJ54428/KIAA0918/KIAA1910/LRRC12/RP11-395N17.1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| SLITRK1 | |
| SLITRK1 | |
| A synthetic peptide corresponding to C-terminus 16 amino acids of human SLITRK1. | |
| RUO | |
| 114798 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only