missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLIT3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLIT3.
Brand: Abnova PAB11260.100ug
This item is not returnable.
View return policy
Specifications
| SLIT3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SLIT3. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| FLJ10764/MEGF5/SLIL2/SLIT1/Slit-3/slit2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Bovine, Canine, Chicken, Chimpanzee, Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| SLIT3 | |
| SLIT3 | |
| A synthetic peptide corresponding to amino acids 1164-1177 of human SLIT3. | |
| RUO | |
| 6586 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction