Learn More
SLC9A3R2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SLC9A3R2.
Brand: Abnova PAB20079.100uL
Description
NHE3 (SLC9A3; MIM 182307) is a sodium/hydrogen exchanger in the brush border membrane of the proximal tubule, small intestine, and colon that plays a major role in transepithelial sodium absorption. SLC9A3R2, as well as SLC9A3R1 (MIM 604990) and protein kinase A phosphorylation, may play a role in NHE3 regulation.[supplied by OMIM
Sequence: RLVEVNGVNVEGETHHQVVQRIKAVEGQTRLLVVDQETDEELRRRQLTCTEEMAQRGLPPAHDPWEPKPDWAHTGSHSSEAGKKDVSGPLRELRPRLCHLRKGPQGYGFNLHSpecifications
| SLC9A3R2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SLC9A3R2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| E3KARP/MGC104639/NHE3RF2/NHERF-2/NHERF2/OCTS2/SIP-1/SIP1/TKA-1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9351 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| SLC9A3R2 | |
| SLC9A3R2 | |
| Recombinant protein corresponding to amino acids of human SLC9A3R2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only