Learn More
SLC6A3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLC6A3.
Brand:  Abnova PAB9597.100uL
Product Code. 16071758
  • £535.00 / 100µL
Description
The dopamine transporter (DAT1) mediates the active reuptake of dopamine from the synapse and is a principal regulator of dopaminergic neurotransmission. The DAT1 gene has been implicated in human disorders such as parkinsonism, Tourette syndrome, and substance abuse (Vandenbergh et al., 1992 [PubMed 1359373]).[supplied by OMIM
Specifications
Specifications
SLC6A3
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of SLC6A3.
In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
DAT/DAT1
Rabbit
Affinity purification
RUO
6531
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Immunohistochemistry (Frozen), Western Blot
Unconjugated
Immunohistochemistry (Frozen sections) (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
SLC6A3
SLC6A3
A synthetic peptide (conjugated with KLH) corresponding to human SLC6A3.