missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SLC6A1.
Brand: Abnova PAB20649.100uL
This item is not returnable.
View return policy
Description
The SLC6A1 gene encodes a gamma-aminobutyric acid (GABA) transporter, which removes GABA from the synaptic cleft (Hirunsatit et al., 2009 [PubMed 19077666]).[supplied by OMIM
Sequence: IYYLYNSFTTTLPWKQCDNPWNTDRCFSNYSMVNTTNMTSAVVEFWERNMHQMTDGLDKPGSpecifications
| SLC6A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SLC6A1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| GABATHG/GABATR/GAT1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6529 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| SLC6A1 | |
| SLC6A1 | |
| Recombinant protein corresponding to amino acids of human SLC6A1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction