Learn More
SLC5A11 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyconal antibody raised against synthetic peptide of SLC5A11.
Brand: Abnova PAB28333.50ug
Description
Cotransporters, such as SLC5A11, represent a major class of proteins that make use of ion gradients to drive active transport for the cellular accumulation of nutrients, neurotransmitters, osmolytes, and ions Roll et al. (2002) [PubMed 12039040].[supplied by OMIM
Specifications
| SLC5A11 | |
| Polyclonal | |
| Rabbit polyconal antibody raised against synthetic peptide of SLC5A11. | |
| In PBS (0.09% sodium azide) | |
| KST1/RKST1/SGLT6/SMIT2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 115584 | |
| Human, Monkey | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/ml) The optimal working dilution should be determined by the end user. | |
| SLC5A11 | |
| SLC5A11 | |
| A synthetic peptide corresponding to 16 amino acids at internal region of human SLC5A11. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only