missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC43A2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SLC43A2.
Brand: Abnova PAB21430.100uL
This item is not returnable.
View return policy
Description
System L amino acid transporters, such as SLC43A2, mediate sodium-independent transport of bulky neutral amino acids across cell membranes (Bodoy et al., 2005 [PubMed 15659399]).[supplied by OMIM
Sequence: YRRQLERQLQQRQEDDKLFLKINGSSNQEAFVSpecifications
| SLC43A2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SLC43A2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ23848/LAT4/MGC34680 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 124935 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| SLC43A2 | |
| SLC43A2 | |
| Recombinant protein corresponding to amino acids of human SLC43A2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction