missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slc3a1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Slc3a1.
Brand: Abnova PAB8505.25ug
This item is not returnable.
View return policy
Specifications
| Slc3a1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Slc3a1. | |
| In PBS (0.1% proclin, 2.0% Block Ace) | |
| Slc3a1 | |
| A synthetic peptide corresponding to rat Slc3a1. | |
| RUO | |
| 29484 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (0.1-0.5 ug/mL) Western Blot (0.01-0.1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Slc3a1 | |
| Rabbit | |
| 25 μg | |
| Primary | |
| Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction