Learn More
SLC39A8 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SLC39A8.
Brand: Abnova PAB23353.100uL
Description
This gene encodes a member of the SLC39 family of solute-carrier genes, which show structural characteristics of zinc transporters. The encoded protein is glycosylated and found in the plasma membrane and mitochondria, and functions in the cellular import of zinc at the onset of inflammation. It is also thought to be the primary transporter of the toxic cation cadmium, which is found in cigarette smoke. Multiple transcript variants encoding different isoforms have been found for this gene. Additional alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. [provided by RefSeq
Sequence: ANLSLSAAQLQHLLEQMGAASRVGVPEPGQLHFNQCLTAEEIFSLHGFSNATQITSSKFSVICPAVLQQLNFHPCEDRPKHKTRPSHSSpecifications
| SLC39A8 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SLC39A8. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| BIGM103/LZT-Hs6/PP3105/ZIP8 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 64116 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| SLC39A8 | |
| SLC39A8 | |
| Recombinant protein corresponding to amino acids of human SLC39A8. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only