missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35D3 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLC35D3.
Brand: Abnova PAB25739.100ug
This item is not returnable.
View return policy
Specifications
| SLC35D3 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunocytochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| SLC35D3 | |
| SLC35D3 | |
| A synthetic peptide corresponding to 17 amino acids at C-terminus of human SLC35D3. | |
| 100 μg | |
| Primary | |
| At least two isoforms of SLC35D3 are known to exist; this antibody will recognize both isoforms. SLC35D3 antibody is predicted to not cross-react with SLC35D1 or SLC35D2. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of SLC35D3. | |
| In PBS (0.02% sodium azide) | |
| FRCL1/MGC102873/bA55K22.3 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 340146 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction