Learn More
SLC35D2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLC35D2.
Brand: Abnova PAB25738.100ug
Description
Nucleotide sugars, which are synthesized in the cytosol or the nucleus, are high-energy donor substrates for glycosyltransferases located in the lumen of the endoplasmic reticulum and Golgi apparatus. Translocation of nucleotide sugars from the cytosol into the lumen compartment is mediated by specific nucleotide sugar transporters, such as SLC35D2 (Suda et al., 2004 [PubMed 15082721]).[supplied by OMIM
Specifications
| SLC35D2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunocytochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| SLC35D2 | |
| SLC35D2 | |
| A synthetic peptide corresponding to 14 amino acids at C-terminus of human SLC35D2. | |
| 100 μg | |
| Primary | |
| At least two isoforms of SLC35D2 are known to exist; this antibody will recognize both isoforms. SLC35D2 antibody is predicted to not cross-react with SLC35D1 or SLC35D3. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of SLC35D2. | |
| In PBS (0.02% sodium azide) | |
| HFRC1/MGC117215/MGC142139/SQV7L/UGTrel8/hfrc | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 11046 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only