Learn More
SLC31A1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SLC31A1.
Brand: Abnova PAB14558.100uL
Description
Copper is an element essential for life, but excessive copper can be toxic or even lethal to the cell. Therefore, cells have developed sophisticated ways to maintain a critical copper balance, with the intake, export, and intracellular compartmentalization or buffering of copper strictly regulated. The 2 related genes ATP7A (MIM 300011) and ATP7B (MIM 606882), responsible for the human diseases Menkes syndrome (MIM 309400) and Wilson disease (MIM 277900), respectively, are involved in copper export. In S. cerevisiae, the copper uptake genes CTR1, CTR2, and CTR3 have been identified, and in human the CTR1 and CTR2 (MIM 603088) genes have been identified.[supplied by OMIM
Specifications
| SLC31A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SLC31A1. | |
| In Tris-citrate/phosphate buffer, pH 7-8 (0.09% sodium azide) | |
| COPT1/CTR1/MGC75487/hCTR1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Pig, Rat, Xenopus species, Zebrafish | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:250) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| SLC31A1 | |
| SLC31A1 | |
| A synthetic peptide corresponding to C-terminus of human SLC31A1. | |
| RUO | |
| 1317 | |
| Store at 4°C. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only