missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC2A9 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Slc2a9.
Brand: Abnova PAB12029.100uL
This item is not returnable.
View return policy
Specifications
| SLC2A9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Slc2a9. | |
| In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
| Glut9 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Mouse | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Slc2a9 | |
| Slc2a9 | |
| A synthetic peptide corresponding to amino acids 400-450 of mouse Slc2a9. | |
| RUO | |
| 117591 | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction