Learn More
SLC28A3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SLC28A3.
Brand: Abnova PAB21556.100uL
Description
Nucleoside transporters, such as SLC28A3, regulate multiple cellular processes, including neurotransmission, vascular tone, adenosine concentration in the vicinity of cell surface receptors, and transport and metabolism of nucleoside drugs. SLC28A3 shows broad specificity for pyrimidine and purine nucleosides (Ritzel et al., 2001 [PubMed 11032837]).[supplied by OMIM
Sequence: MELRSTAAPRAEGYSNVGFQNEENFLENENTSGNNSIRSRAVQSREHTNTKQDEEQVTVEQDSPRNREHMEDDDEEMQQKGCLERRYDTVCGFCRKHKTTLRHSpecifications
| SLC28A3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SLC28A3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CNT3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 64078 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| SLC28A3 | |
| SLC28A3 | |
| Recombinant protein corresponding to amino acids of human SLC28A3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only