missing translation for 'onlineSavingsMsg'
Learn More

SLC26A1, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant SLC26A1.

Brand:  Abnova PAB23838.100uL

Product Code. 16186250

  • £535.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene is a member of a family of sulfate/anion transporter genes. Family members are well conserved in their genomic (number and size of exons) and protein (aa length among species) structures, but have markedly different tissue expression patterns. This gene is primarily expressed in the liver, pancreas, and brain. Three splice variants that encode different isoforms have been identified. [provided by RefSeq

Sequence: DTAFYEDATEFEGLVPEPGVRVFRFGGPLYYANKDFFLRSLYSLTGLDAGCMAARRKEGGSETGVGEGGP
Specifications

Specifications

SLC26A1
Polyclonal
Rabbit polyclonal antibody raised against recombinant SLC26A1.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
EDM4/SAT-1/SAT1
Rabbit
Antigen affinity purification
RUO
10861
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:1000-1:2500) The optimal working dilution should be determined by the end user.
SLC26A1
SLC26A1
Recombinant protein corresponding to amino acids of human SLC26A1.
100 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only