Learn More
SLC22A8 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SLC22A8.
Brand: Abnova PAB28282.100uL
Description
The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. [provided by RefSeq
Sequence: KKEEGERLSLEELKLNLQKEISLAKAKYTASDLFRIPMLRSpecifications
| SLC22A8 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SLC22A8. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| MGC24086/OAT3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9376 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin) | |
| Unconjugated | |
| Immunohistochemistry (1:2500-1:5000) The optimal working dilution should be determined by the end user. | |
| SLC22A8 | |
| SLC22A8 | |
| Recombinant protein corresponding to amino acids of recombinant SLC22A8. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only