missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC12A3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Human, Rat SLC12A3.
Brand: Abnova PAB28748.100ug
This item is not returnable.
View return policy
Specifications
| SLC12A3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant Human, Rat SLC12A3. | |
| PBS, 50% glycerol and 0.09% sodium azide | |
| SLC12A3 | |
| Recombinant protein corresponding to SLC12A3 N-terminus (amino acids 74-95). | |
| 100 μg | |
| Primary | |
| 1 ug/mL of PAB28748 was sufficient for detection of SLC12A3 ,∼160kDa, in 10ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Immunofluorescence Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| FLJ96318/NCCT/TSC | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 6559/54300 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction