Learn More
SKAP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SKAP1.
Brand: Abnova PAB28622.100uL
Description
This gene encodes a T cell adaptor protein, a class of intracellular molecules with modular domains capable of recruiting additional proteins but that exhibit no intrinsic enzymatic activity. The encoded protein contains a unique N-terminal region followed by a PH domain and C-terminal SH3 domain. Along with the adhesion and degranulation-promoting adaptor protein, the encoded protein plays a critical role in inside-out signaling by coupling T-cell antigen receptor stimulation to the activation of integrins. [provided by RefSeq
Sequence: SLTIPYEEDEEEEEKEETYDDIDGFDSPSCGSQCRPTILPGSVGIKEPTEEKEEEDIYEVLPDEEHDLEEDESGTRRKGVDYASYYQGLWDCHGDQPDELSFQRGDLSpecifications
| SKAP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SKAP1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| SCAP1/SKAP55 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8631 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| SKAP1 | |
| SKAP1 | |
| Recombinant protein corresponding to amino acids of human SKAP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only