Learn More
SIGMAR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SIGMAR1.
Brand: Abnova PAB20993.100uL
Description
This gene encodes a receptor protein that interacts with a variety of psychotomimetic drugs, including cocaine and amphetamines. The receptor is believed to play an important role in the cellular functions of various tissues associated with the endocrine, immune, and nervous systems. As indicated by its previous name, opioid receptor sigma 1 (OPRS1), the product of this gene was erroneously thought to function as an opioid receptor; it is now thought to be a nonopioid receptor. Alternative splicing of this gene results in transcript variants encoding distinct isoforms. [provided by RefSeq
Sequence: FVFQREEIAQLARQYAGLDHELAFSRLIVELRRLHPGHVLPDEELQWVFVNAGGSpecifications
| SIGMAR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SIGMAR1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ25585/MGC3851/OPRS1/SR-BP1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10280 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| SIGMAR1 | |
| SIGMAR1 | |
| Recombinant protein corresponding to amino acids of human SIGMAR1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only