missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Sheep IgM (H+L) Secondary Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68364
This item is not returnable.
View return policy
Description
IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Sheep samples. It is validated for Flow Cytometry
Specifications
| IgM (H+L) | |
| Polyclonal | |
| FITC | |
| PBS, 0.2% BSA | |
| Sheep IgM | |
| 1 mg | |
| B Cell Development and Differentiation Markers, Immunology | |
| Sheep | |
| Purified |
| Flow Cytometry | |
| 1 mg/mL | |
| Flow Cytometry 1:200 - 1:2000 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction