missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Sheep IgG (H+L) Secondary Antibody, FITC

Product Code. 11879180
Change view
Click to view available options
Quantity:
1.5 mg
Unit Size:
1.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11879180 1.5 mg 1.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11879180 Supplier Invitrogen™ Supplier No. 31627
  • ONLINE EXCLUSIVE PRICE
    £137.00
    £110.97 / 1.5mg
    Save £26.03
    19% Off
View all promotions
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31627 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31627 reacts with whole molecule sheep IgG. It also reacts with the light chains of other sheep immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Fluorophore: Fluorescein-5-isothiocyanate (FITC-isomer 1) Amax = 492 nm; Emax = 520 nm Fluorophore/Protein: 9.1 μg/mg; 3.4 moles FITC per mole IgG Reconstitute with 1.1 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA.

Anti-Sheep secondary antibodies are affinity-purified antibodies with well-characterized specificity for ovine (sheep) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sheep IgG (H+L)
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate FITC
Formulation PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Host Species Rabbit
Purification Method Antigen affinity chromatography
Quantity 1.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Sheep
Content And Storage 4°C, store in dark
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
compliance-icons
Product Identifier
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, FITC
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.