Learn More
Rabbit anti-Sheep IgG - H&L (HRP), Secondary Antibody, Abcam™
Brand: Abcam AB6747-1MG
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Sheep
Immunogen: Sheep IgG whole molecule
Applications:
Dot Blotting, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blotting
Specifications
| Sheep IgG - H&L (HRP) | |
| 2mg/mL | |
| Rabbit | |
| 1 mg | |
| Secondary | |
| IgG |
| Polyclonal | |
| HRP | |
| Affinity Purified | |
| Immunology | |
| Sheep |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.