missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sgo1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Sgo1.
Brand: Abnova PAB15588.100uL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
| Sgo1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant Sgo1. | |
| In antiserum (0.05% sodium azide) | |
| sgo1 | |
| Recombinant protein corresponding to Schizosaccharomyces pombe Sgo1. | |
| RUO | |
| 2541326 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunofluorescence (1:100) The optimal working dilution should be determined by the end user. | |
| sgo1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| S. pombe | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction