Learn More
SGCZ, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SGCZ.
Brand: Abnova PAB20947.100uL
Description
The zeta-sarcoglycan gene measures over 465kb and localizes to 8p22. This protein is part of the sarcoglycan complex, a group of 6 proteins. The sarcoglycans are all N-glycosylated transmembrane proteins with a short intra-cellular domain, a single transmembrane region and a large extra-cellular domain containing a carboxyl-terminal cluster with several conserved cysteine residues. The sarcoglycan complex is part of the dystrophin-associated glycoprotein complex (DGC), which bridges the inner cytoskeleton and the extra-cellular matrix. [provided by RefSeq
Sequence: PLYVKEIHSRKDSPLVLQSDRNVTVNARNHMGQLTGQLTIGADAVEAQCKRFEVRASEDGRVLFSADEDEITIGAEKLKVTGTEGAVFGHSVETPHIRAEPSQDLSpecifications
| SGCZ | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SGCZ. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MGC149397/ZSG1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 137868 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| SGCZ | |
| SGCZ | |
| Recombinant protein corresponding to amino acids of human SGCZ. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only