missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFXN4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SFXN4.
Brand: Abnova PAB17399.100ug
This item is not returnable.
View return policy
Specifications
| SFXN4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SFXN4. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| BCRM1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total SFXN4 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| SFXN4 | |
| SFXN4 | |
| A synthetic peptide corresponding to N-terminus of human SFXN4. | |
| RUO | |
| 119559 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction