missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sftpa1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Sftpa1.
Brand: Abnova PAB19512.100ug
This item is not returnable.
View return policy
Specifications
| Sftpa1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Sftpa1. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| SP-A/Sftp-1/Sftp1 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 20387 | |
| Mouse | |
| Lyophilized |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Sftpa1 | |
| Sftpa1 | |
| A synthetic peptide corresponding to amino acids at C-terminus of mouse Sftpa1. | |
| 100 μg | |
| Primary | |
| It identical to the related rat sequence. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction