missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SF4.
Brand: Abnova PAB20289.100uL
This item is not returnable.
View return policy
Description
SF4 is a member of the SURP family of splicing factors.[supplied by OMIM
Sequence: GKANRWFGVAPPKSGKMNMNILHQEELIAQKKREIEAKMEQKAKQNQVASPQPPHPGEITNAHNSSCISNKFANDGSFLQQFLKLQKAQTSTDAPTSAPSAPPSTPTSpecifications
| SF4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SF4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp434E2216/F23858/RBP | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57794 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| SF4 | |
| SF4 | |
| Recombinant protein corresponding to amino acids of human SF4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction