Learn More
SF3B14 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SF3B14.
Brand: Abnova PAB22832.100uL
Description
This gene encodes a 14kDa protein subunit of the splicing factor 3b complex. Splicing factor 3b associates with both the U2 and U11/U12 small nuclear ribonucleoprotein complexes (U2 snRNP) of spliceosomes. This 14kDa protein interacts directly with subunit 1 of the splicing factor 3b complex. This 14kDa protein also interacts directly with the adenosine that carries out the first transesterification step of splicing at the pre-mRNA branch site. [provided by RefSeq
Sequence: MAMQAAKRANIRLPPEVNRILYIRNLPYKITAEEMYDIFGKYGPIRQIRVGNTPETRGTAYVVSpecifications
| SF3B14 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SF3B14. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CGI-110/HSPC175/Ht006/P14/SAP14/SF3B14a | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51639 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| SF3B14 | |
| SF3B14 | |
| Recombinant protein corresponding to amino acids of human SF3B14. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only