Learn More
SETDB1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SETDB1.
Brand: Abnova PAB10044.100ug
Description
This gene encodes a histone methyltransferase. The encoded enzyme catalyzes the reaction of S-adenosyl-L-methionine and histone L-lysine to produce S-adenosyl-L-homocysteine and histone N(6)-methyl-L-lysine. The encoded protein likely functions in transcriptional repression. Alternatively spliced transcript variants have been described
Specifications
| SETDB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SETDB1. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| ESET/KG1T/KIAA0067/KMT1E | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| ELISA (1:10000-1:50000) Western Blot (1:1000-1:2500) The optimal working dilution should be determined by the end user. | |
| SETDB1 | |
| SETDB1 | |
| A synthetic peptide corresponding to amino acids 1058-1075 of human SETDB1. | |
| RUO | |
| 9869 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only